Learn More
Analytical Sales & Services HotSleeve+ Column Heater, Self-Calibrating, Flexible Heating Unit, Units: Each
Catalog No. NC3596829
Encompass
The HotSleeve+ Column Heater is a self-calibrating heating unit engineered for precise temperature control of HPLC columns. With a flexible design and a temperature range from 5ºC above ambient to 85ºC, it enhances performance for scientific applications. Compatible with MultiSLEEVE™ or MonoSLEEVE™ controllers (sold separately), it offers reliable temperature management, setting it apart from other heating units.
Catalog No. NC3596829 Supplier Analytical Sales & Services Supplier No. HSI0806L
The HotSleeve+ Column Heater is a self-calibrating heating unit engineered for precise temperature control of HPLC columns. With a flexible design and a temperature range from 5ºC above ambient to 85ºC, it enhances performance for scientific applications. Compatible with MultiSLEEVE™ or MonoSLEEVE™ controllers (sold separately), it offers reliable temperature management, setting it apart from other heating units.