Learn More
Analytical Sales & Services HotSleeve+ Column Heater - 0.8" x 14"
Catalog No. 5000805240
Encompass
HotSleeve+ Self-calibrating Heater for HPLC Columns\n\nThe HotSleeve+ replaces the original Hot Sleeve column heater. Use with either the MultiSLEEVE™ controller or MonoSLEEVE™ controller, sold separately.\n\n• Inner diameter: 0.8"\n• Length: 14"\n• Temperature range:  5ºC Above Ambient to 85ºC
Catalog No. 50-008-05240 Supplier Analytical Sales & Services Supplier No. HSI0814L
HotSleeve+ Self-calibrating Heater for HPLC Columns\n\nThe HotSleeve+ replaces the original Hot Sleeve column heater. Use with either the MultiSLEEVE™ controller or MonoSLEEVE™ controller, sold separately.\n\n• Inner diameter: 0.8"\n• Length: 14"\n• Temperature range:  5ºC Above Ambient to 85ºC