Learn More
Analytical Sales & Services HotSleeve+ Column Heater - 2" x 12"
Catalog No. 5000805241
Encompass
HotSleeve+ Self-calibrating Heater for HPLC Columns\n\nThe HotSleeve+ replaces the original Hot Sleeve column heater. Use with either the MultiSLEEVE™ controller or MonoSLEEVE™ controller, sold separately.\n\n• Inner diameter: 2"\n• Length: 12"\n• Temperature range:  5ºC Above Ambient to 85ºC
Catalog No. 50-008-05241 Supplier Analytical Sales & Services Supplier No. HSI2012L
HotSleeve+ Self-calibrating Heater for HPLC Columns\n\nThe HotSleeve+ replaces the original Hot Sleeve column heater. Use with either the MultiSLEEVE™ controller or MonoSLEEVE™ controller, sold separately.\n\n• Inner diameter: 2"\n• Length: 12"\n• Temperature range:  5ºC Above Ambient to 85ºC