Learn More
Analytical Sales & Services HotSleeve+ Column Heater - 3.5" x 12"
Catalog No. 5000805175
Encompass
HotSleeve+ Self-calibrating Heater for HPLC Columns\n\nThe HotSleeve+ replaces the original Hot Sleeve column heater. Use with either the MultiSLEEVE™ controller or MonoSLEEVE™ controller, sold separately.\n\n• Inner diameter: 3.5"\n• Length: 12"\n• Temperature range: 5ºC Above Ambient to 85ºC
Catalog No. 50-008-05175 Supplier Analytical Sales & Services Supplier No. HSI3512L
HotSleeve+ Self-calibrating Heater for HPLC Columns\n\nThe HotSleeve+ replaces the original Hot Sleeve column heater. Use with either the MultiSLEEVE™ controller or MonoSLEEVE™ controller, sold separately.\n\n• Inner diameter: 3.5"\n• Length: 12"\n• Temperature range: 5ºC Above Ambient to 85ºC