Learn More
Roche Diagnostics KAPA2G Fast Multiplex Kits, 500 x 25 uL reactions, 6.25 mL
Catalog No. 501965222 Shop All Roche Diagnostics Products
Encompass
KAPA2G Fast Multiplex PCR Kits contain a second-generation (2G) enzyme derived through our directed evolution technology. KAPA2G FAST HotStart DNA Polymerase is an antibody-mediated hot start formulation engineered for higher processivity and speed, offering significantly faster extension rates than wild-type Taq DNA polymerase. In addition to speed, KAPA2G Fast Multiplex Kit provides higher yields and sensitivity than competitor enzymes, allowing for more uniform multiplexed PCR. *This item is non returnable.
This item is not returnable. View return policy
Catalog No. 50-196-5222 Supplier Roche Diagnostics Supplier No. 07961430001
KAPA2G Fast Multiplex PCR Kits contain a second-generation (2G) enzyme derived through our directed evolution technology. KAPA2G FAST HotStart DNA Polymerase is an antibody-mediated hot start formulation engineered for higher processivity and speed, offering significantly faster extension rates than wild-type Taq DNA polymerase. In addition to speed, KAPA2G Fast Multiplex Kit provides higher yields and sensitivity than competitor enzymes, allowing for more uniform multiplexed PCR. *This item is non returnable.