Learn More
LAMDA BIOTECH INC HotStart PCR 2x MasterMxGreen
Catalog No. NC3724377
Encompass_Preferred
A pre-formulated PCR master mix containing chemically modified HotStart Taq DNA polymerase with integrated green tracking dye for direct gel loading capability.
Catalog No. NC3724377 Supplier LAMDA BIOTECH INC Supplier No. D136G
A pre-formulated PCR master mix containing chemically modified HotStart Taq DNA polymerase with integrated green tracking dye for direct gel loading capability.