Learn More
LAMDA BIOTECH INC HotStart Taq DNA Polymerase
Supplier:  LAMDA BIOTECH INC D129500
Encompass_Preferred
The HotStart Taq DNA Polymerase is a chemically modified Taq DNA Polymerase, whose enzyme activities can only be activated after 3-5 minutes of incubation at 94o C. The HotStart Taq Polymerase uses amplification conditions for regular Taq DNA Polymerase, except no polymerase activity will be present before the onset of thermal cycling. This prevents nonspecific DNA amplification and primer dimer formation. The amplified products, up to 6Kb in length, contain single 3’-overhanging-A ends. This allows for TA cloning methods, if the amplified fragments need to be cloned.
Catalog No. 50-204-5768