Learn More
Apexbio Technology LLC HOTSTART UNIV BLUE 2X QPCR-5mg
SDP
Catalog No. 5000897447
Encompass_Preferred
HotStart Universal Blue 2X Green qPCR Master Mix (with UDG) is a highly efficient reagent used in the field of molecular biology research particularly for quantitative PCR (qPCR) applications Originating from advancements in nucleic acid amplification techniques this master mix enables precise quantification by incorporating a unique blend of components including a UDG enzyme that prevents carryover contamination Its mechanism of action involves the hot-start activation of polymerase ensuring high specificity and sensitivity The mix demonstrates robust in vitro activity across a range of reactions typically seen in nanomolar to low micromolar concentrations It is commonly employed in a variety of experimental setups including cell models and high-throughput drug screening assays Experimental concentrations typically depend on the specific research design offering versatility across myriad applications in genetic analysis gene expression studies and diagnostic research
Catalog No. 50-008-97447 Supplier Apexbio Technology LLC Supplier No. K15465
HotStart Universal Blue 2X Green qPCR Master Mix (with UDG) is a highly efficient reagent used in the field of molecular biology research particularly for quantitative PCR (qPCR) applications Originating from advancements in nucleic acid amplification techniques this master mix enables precise quantification by incorporating a unique blend of components including a UDG enzyme that prevents carryover contamination Its mechanism of action involves the hot-start activation of polymerase ensuring high specificity and sensitivity The mix demonstrates robust in vitro activity across a range of reactions typically seen in nanomolar to low micromolar concentrations It is commonly employed in a variety of experimental setups including cell models and high-throughput drug screening assays Experimental concentrations typically depend on the specific research design offering versatility across myriad applications in genetic analysis gene expression studies and diagnostic research