Learn More
Cole-Parmer Oakton by Cole-Parmer Threaded In-Line Housing with Guard
Supplier:  Cole-Parmer 0599377
Encompass_Preferred
Simply slip an electrode into this housing and screw it into a tee fitting. Its pipe tee connection is 1/2 inch NPT(M).
Catalog No. 50-633-435