Learn More
Cole-Parmer Cole-Parmer pH Electrode Housing, threaded/in-line, w/o bulb guard, PVDF
Catalog No. 50633434 Shop All Cole-Parmer Products
Encompass_Preferred
Simply slip an electrode into this housing and screw it into a tee fitting. Its pipe tee connection is 1/2 inch NPT(M).
Catalog No. 50-633-434 Supplier Cole-Parmer Supplier No. 0599376
Simply slip an electrode into this housing and screw it into a tee fitting. Its pipe tee connection is 1/2 inch NPT(M).