Learn More
Science First™ How Clean is the Air?
SDPSCI_ED
Catalog No. S25163 Shop All Science First Products
Change view
Click to view available options
Product Type:
How Clean Is the Air Kit
Catalog No. Product Type
S25163 How Clean Is the Air Kit
1 options
Catalog No. S25163 Supplier Science First™ Supplier No. 5600
Add to Cart
Add to Cart
Through a variety of laboratory exercises, students will monitor air particles and explore the impact and effects of air pollutants on materials and on the chemistry of rainwater.
Each kit contains: two metal cups, one magnifying glass, one plastic syringe, one plastic cup, one test tube, one pipette, two wooden mixers, six labels, two straws, limestone, vinegar, nitrogen oxide test fabric, bromothymol blue solution on mesh, and a teacher's guide.
Specifications
For Use With (Application) To demonstrate the effects of pollutants on rain water chemistry
Includes Two metal cups, one magnifying glass, one plastic syringe, one plastic cup, one test tube, one pipette, two wooden mixers, six labels, two straws, limestone, vinegar, nitrogen oxide test fabric, bromothymol blue solution on mesh, and a teacher guide
Product Type How Clean Is the Air Kit