Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ How Clean is Clean? Testing the Effectiveness of Antibacterial Cleaners Kit
SDPSCI_ED

Catalog No. S27556 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27556 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27556 Supplier Edvotek™ Supplier No. 161
Add to Cart
Add to Cart

Students will do two experiments to explore the properties of bacterial growth. They will determine a bacteria's ability to resist the antibiotic ampicillin, then test different household cleaners to determine which one is the most effective at preventing bacterial growth.

  • Storage: Some Components Require Refrigerator Storage Upon Receipt
  • Complete in 2 Lab Periods (24 Hours)
  • For 10 Groups

Specifications

Grade 9 to Undergraduate
Age 14 to 20
Includes Ampicillin, E. coli BactoBeads™, ReadyPour™ Agar (sterile), Recovery Broth, Petri Plates (Small & Large), Plastic Microtipped Transfer Pipettes,10 mL Pipette (wrapped, sterile), Swabs (sterile), Microcentrifuge Tubes, and Disks for Antibiotics
Requires Microwave, Adjustable Volume Micropipette (50-50 μL and 50-200 μL), Incubation Oven (37°C), Pipette Pumps/Bulbs, Marking Pens, Gloves, Safety Goggles, Variety of Household Cleaners
Time Range 2 Lab Periods (24 Hr.)
Content And Storage Some Components Require Refrigerator Storage Upon Receipt
Class Size 10 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.