Learn More
MSC Howard Leight Earmuff, 23 dB NRR, Over Head, Folding, Black
Catalog No. 5000509005
Encompass
These earmuffs offer reliable hearing protection, reducing noise across all frequencies with patented Air Flow Control Technology. Their ultra-lightweight, compact folding design ensures convenient storage and portability. Designed for comfort, they feature exterior foam padding and super-soft ear cushions to prevent discomfort, and snap-in ear cushions allow for easy cleaning.
  • Sleek, low-profile earcup design
  • Ultra-lightweight
  • Compact folding for storage
  • Padded, adjustable headband
  • 23 dB noise reduction rating
  • Black cup color
  • One size fits most
  • Patented Air Flow Control Technology
  • Exterior foam padding and soft ear cushions
  • Snap-in ear cushions for easy cleaning
  • Over-head band position
  • Non-conductive
Catalog No. 50-005-09005 Supplier MSC Supplier No. 31310600
These earmuffs offer reliable hearing protection, reducing noise across all frequencies with patented Air Flow Control Technology. Their ultra-lightweight, compact folding design ensures convenient storage and portability. Designed for comfort, they feature exterior foam padding and super-soft ear cushions to prevent discomfort, and snap-in ear cushions allow for easy cleaning.
  • Sleek, low-profile earcup design
  • Ultra-lightweight
  • Compact folding for storage
  • Padded, adjustable headband
  • 23 dB noise reduction rating
  • Black cup color
  • One size fits most
  • Patented Air Flow Control Technology
  • Exterior foam padding and soft ear cushions
  • Snap-in ear cushions for easy cleaning
  • Over-head band position
  • Non-conductive