Learn More
MSC Howard Leight Earmuffs, 26 dB NRR, Over Head, Gray
Catalog No. 5000509109
Encompass
The Howard Leight VeriShield VS120 Series Passive Earmuffs provide reliable hearing protection for workers in demanding environments. Designed for extended wear, these earmuffs combine a comfortable fit with effective noise reduction, making them suitable for construction, manufacturing, and industrial settings.
  • Delivers 26 dB noise reduction.
  • Wide cup opening accommodates larger ears or hearing aids.
  • Cushioned and adjustable headband reduces pressure and enhances comfort.
  • Memory foam ear cushions provide greater comfort and attenuation.
  • Robust, oil-resistant construction withstands heavy-work environments.
  • Air Flow Control technology provides optimum attenuation across all frequencies.
  • Designed for seamless integration with other personal protective equipment.
Catalog No. 50-005-09109 Supplier MSC Supplier No. 95460507
The Howard Leight VeriShield VS120 Series Passive Earmuffs provide reliable hearing protection for workers in demanding environments. Designed for extended wear, these earmuffs combine a comfortable fit with effective noise reduction, making them suitable for construction, manufacturing, and industrial settings.
  • Delivers 26 dB noise reduction.
  • Wide cup opening accommodates larger ears or hearing aids.
  • Cushioned and adjustable headband reduces pressure and enhances comfort.
  • Memory foam ear cushions provide greater comfort and attenuation.
  • Robust, oil-resistant construction withstands heavy-work environments.
  • Air Flow Control technology provides optimum attenuation across all frequencies.
  • Designed for seamless integration with other personal protective equipment.