Learn More
MSC Howard Leight Earmuffs, Hard Hat Capmount, 27 dB NRR, Hi-Viz Yellow
Catalog No. 5000509258
Encompass
These earmuffs provide superior NRR 27 hearing protection with a convenient hard hat attachment. Designed for optimum comfort during long periods of use, they are lightweight and accommodate a wide range of head and ear sizes, featuring memory foam ear cushions and an adjustable design for an improved fit.
  • Delivers superior 27 dB NRR hearing protection.
  • Features convenient hard hat attachment.
  • Designed with a lightweight construction.
  • Provides improved comfort, fit, and function.
  • Accommodates a wide range of head and ear sizes.
  • Incorporates memory foam ear cushions for enhanced comfort and attenuation.
  • Equipped with an oil-resistant cup surface and ABS molded cups for durability in demanding environments.
  • Utilizes Air Flow Control Technology for optimal attenuation across all frequencies.
  • Hi-viz yellow color provides added safety.
Catalog No. 50-005-09258 Supplier MSC Supplier No. 95460531
These earmuffs provide superior NRR 27 hearing protection with a convenient hard hat attachment. Designed for optimum comfort during long periods of use, they are lightweight and accommodate a wide range of head and ear sizes, featuring memory foam ear cushions and an adjustable design for an improved fit.
  • Delivers superior 27 dB NRR hearing protection.
  • Features convenient hard hat attachment.
  • Designed with a lightweight construction.
  • Provides improved comfort, fit, and function.
  • Accommodates a wide range of head and ear sizes.
  • Incorporates memory foam ear cushions for enhanced comfort and attenuation.
  • Equipped with an oil-resistant cup surface and ABS molded cups for durability in demanding environments.
  • Utilizes Air Flow Control Technology for optimal attenuation across all frequencies.
  • Hi-viz yellow color provides added safety.