Learn More
MSC Howard Leight Earmuffs, Passive, NRR 30 dB
Catalog No. 5000508326
Encompass
These earmuffs deliver high performance and robust steel wire durability, ensuring comfort during daily use. They feature patented Air Flow Control technology for optimal sound attenuation across all frequencies. The sleek, contemporary design integrates well with other personal protective equipment, providing maximum protection.
  • Noise reduction rating (NRR) of 30 dB
  • Over-the-head, noise-blocking design
  • Super-soft, snap-in ear cushions for easy replacement and comfort
  • Telescopic, steel wire headband for adjustable, secure fit
  • Padded foam headband minimizes pressure
  • Patented Air Flow Control technology for consistent sound reduction
  • Meets ANSI S3.19-1974 Standards
  • Compatible with other PPE such as faceshields and hardhats
Catalog No. 50-005-08326 Supplier MSC Supplier No. 31310527
These earmuffs deliver high performance and robust steel wire durability, ensuring comfort during daily use. They feature patented Air Flow Control technology for optimal sound attenuation across all frequencies. The sleek, contemporary design integrates well with other personal protective equipment, providing maximum protection.
  • Noise reduction rating (NRR) of 30 dB
  • Over-the-head, noise-blocking design
  • Super-soft, snap-in ear cushions for easy replacement and comfort
  • Telescopic, steel wire headband for adjustable, secure fit
  • Padded foam headband minimizes pressure
  • Patented Air Flow Control technology for consistent sound reduction
  • Meets ANSI S3.19-1974 Standards
  • Compatible with other PPE such as faceshields and hardhats