Learn More
MSC Howard Leight Earmuffs, Plastic, 27 dB, Over Head, Folding
Catalog No. 5000507828
Encompass
These durable earmuffs provide a 27 dB noise reduction rating, designed for daily use and comfort. Featuring robust steel-wire construction and a padded foam headband, they offer optimal attenuation and easy maintenance.
  • Over head band position
  • Cushioned and adjustable headband
  • Folding design
  • Non-conductive
  • Snap-in ear cushions for easy maintenance
  • Optimal attenuation across all frequencies
Catalog No. 50-005-07828 Supplier MSC Supplier No. 31310584
These durable earmuffs provide a 27 dB noise reduction rating, designed for daily use and comfort. Featuring robust steel-wire construction and a padded foam headband, they offer optimal attenuation and easy maintenance.
  • Over head band position
  • Cushioned and adjustable headband
  • Folding design
  • Non-conductive
  • Snap-in ear cushions for easy maintenance
  • Optimal attenuation across all frequencies