Learn More
MSC Howard Leight Earplugs, 28 dB NRR, Foam, Push-In, Uncorded
Catalog No. 5000508601
Encompass
These earplugs provide 28 dB noise reduction and are made from foam with a push-in insertion method. They are uncorded and reusable, suitable for various environments.
  • Reusable design for extended use.
  • Uncorded attachment for convenience.
  • Pod shape for comfortable fit.
  • Foam material for effective noise blocking.
  • Push-in insertion method for easy application.
  • Universal size fits most users.
  • Packaged in a quantity of 100 pairs.
  • Non-metal detectable for specific environments.
  • Features closed cell foam.
Catalog No. 50-005-08601 Supplier MSC Supplier No. 42436204
These earplugs provide 28 dB noise reduction and are made from foam with a push-in insertion method. They are uncorded and reusable, suitable for various environments.
  • Reusable design for extended use.
  • Uncorded attachment for convenience.
  • Pod shape for comfortable fit.
  • Foam material for effective noise blocking.
  • Push-in insertion method for easy application.
  • Universal size fits most users.
  • Packaged in a quantity of 100 pairs.
  • Non-metal detectable for specific environments.
  • Features closed cell foam.