Learn More
MSC Howard Leight Electronic Earmuff, 22 dB NRR, Amplifies 82 dB, Folding, Black
Catalog No. 5000508731
Encompass
This electronic earmuff protects wearers from hazardous noise while allowing connection to their environment. It uses built-in directional microphones to amplify ambient sounds to a safe 82 dB, enhancing communication and natural listening. For protection, it actively compresses amplification when impact sound reaches 82 dB, and passively blocks noise at 22 dB NRR.
  • Amplifies low-level sounds up to 4 times for improved awareness.
  • Offers 22 dB NRR for hearing protection.
  • Folding design for easy storage.
  • Aux input for external audio devices.
  • 4-hour automatic shut-off to save battery.
  • Patented Air Flow Control™ Technology for optimal attenuation.
  • Ultraslim ear cups clear firearm stocks.
  • Powered by two included AAA batteries.
Catalog No. 50-005-08731 Supplier MSC Supplier No. 41317009
This electronic earmuff protects wearers from hazardous noise while allowing connection to their environment. It uses built-in directional microphones to amplify ambient sounds to a safe 82 dB, enhancing communication and natural listening. For protection, it actively compresses amplification when impact sound reaches 82 dB, and passively blocks noise at 22 dB NRR.
  • Amplifies low-level sounds up to 4 times for improved awareness.
  • Offers 22 dB NRR for hearing protection.
  • Folding design for easy storage.
  • Aux input for external audio devices.
  • 4-hour automatic shut-off to save battery.
  • Patented Air Flow Control™ Technology for optimal attenuation.
  • Ultraslim ear cups clear firearm stocks.
  • Powered by two included AAA batteries.