Learn More
Honeywell™ Howard Leight™ Leightning L1 Ear Muffs
Catalog No. 19161270
Change view
Click to view available options
L1N; NRR 25dB
Catalog No.
19-161-270 L1N; NRR 25dB
1 options
Catalog No. 19-161-270 Supplier Honeywell™ Supplier No. 1011994
High performance and robust steel wire durability. This noise-blocking earmuff meets NRR 25 Canada Class B requirements.
  • Robust steel wire construction withstands demanding use, especially in tough environments
  • Features patented Air Flow Control technology, delivering optimal attenuation across all frequencies
  • Snap-in ear cushions make replacement quick and easy
  • Multiple attenuation levels allow for targeted attenuation for a variety of applications
  • Telescopic height adjustment remains fixed during use
  • Delivers high degree of comfort; ideal for situations where compact earmuffs and reliable Noise-Blocking protection are required
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Includes Elastic headband strap
Certifications/Compliance ANSI S3.19-1974
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
Noise Reduction Rating (NRR) 25 dB
Product Type Ear Muffs
Dielectric No
Style Behind-the-neck