Learn More
MSC Howard Leight Passive Earmuff, 30 dB NRR, Over the Head, Black
Catalog No. 5000507901
Encompass
This passive earmuff offers superior hearing protection with a 30 dB Noise Reduction Rating. Its lightweight, streamlined design ensures comfort and style, featuring super-soft ear cushions and a telescopic, steel-wire headband for a custom, durable fit. Patented Air Flow Control technology reduces sound across all frequencies.
  • 30 dB Noise Reduction Rating for effective sound reduction.
  • Lightweight, streamlined design for comfort and style.
  • Super-soft ear cushions for prolonged comfort.
  • Snap-in ear cushions for quick replacement and maintenance.
  • Telescopic, steel-wire headband for a custom, durable fit.
  • Padded foam headband to minimize pressure.
  • Air Flow Control technology reduces sound across all frequencies.
  • Steel wire construction for durability.
  • Three sound reduction levels to suit different shooting environments.
Catalog No. 50-005-07901 Supplier MSC Supplier No. 31310485
This passive earmuff offers superior hearing protection with a 30 dB Noise Reduction Rating. Its lightweight, streamlined design ensures comfort and style, featuring super-soft ear cushions and a telescopic, steel-wire headband for a custom, durable fit. Patented Air Flow Control technology reduces sound across all frequencies.
  • 30 dB Noise Reduction Rating for effective sound reduction.
  • Lightweight, streamlined design for comfort and style.
  • Super-soft ear cushions for prolonged comfort.
  • Snap-in ear cushions for quick replacement and maintenance.
  • Telescopic, steel-wire headband for a custom, durable fit.
  • Padded foam headband to minimize pressure.
  • Air Flow Control technology reduces sound across all frequencies.
  • Steel wire construction for durability.
  • Three sound reduction levels to suit different shooting environments.