Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Howard Leight™ Thunder T2 Ear Muffs

Catalog No. 19167850
Change view
Click to view available options
T2H; Cap mount
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-167-850 T2H; Cap mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-167-850 Supplier Honeywell™ Supplier No. 1011602

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Top-of-the-line protection and comfort with dielectric construction. This helmet earmuff meets NRR 25 Canada Class A(L) requirements.

  • Dielectric construction suitable for all workplaces, especially electrical environments
  • Uniform headband pressure for all head sizes, providing better comfort for long-term wear
  • Inner-ventilated headband minimizes pressure on the head, breathes easier in warm/humid climates
  • Non-deforming outer headband withstands rough treatment in the toughest workplaces
  • Patented Air Flow Control technology provides optimal attenuation across all frequencies
  • Quick-Click height adjustment remains fixed during wear
  • Snap-in ear cushions make replacement quick and easy
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Certifications/Compliance ANSI S3.19-1974
For Use With (Equipment) For hard hats
For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
Noise Reduction Rating (NRR) 28 dB
Product Type Thunder T2 Ear Muff
Dielectric Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.