Learn More
Electron Microscopy Sciences Howard Mold Counter with Two Cover Slips
SDP
Catalog No. 5027819
Encompass_Preferred
Howard mold counting chambers is used in making mold counts of tomato products.
Catalog No. 50-278-19 Supplier Electron Microscopy Sciences Supplier No. 6351255
Howard mold counting chambers is used in making mold counts of tomato products.