Learn More
Aviva Systems Biology Corp HOXA10 Antibody - N-terminal region (P100932_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100932_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXA10. Alternate names: PL, HOX1, HOX1H, HOX1.8ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-53807