Learn More
Aviva Systems Biology Corp Hoxa10 Antibody - C-terminal region (ARP58160_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58160_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hoxa10. It was validated on Western Blot. . Alternate names: MGC188427, RGD1564605, Hoxa10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-446