Learn More
Aviva Systems Biology Corp HOXA10 Antibody - middle region (P100768_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100768_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXA10. Alternate names: HOX1, HOX1.8, HOX1H, MGC12859, PLApplicationWB. Species reactivity: Dog, Mouse, Rabbit
Catalog No. 50-014-62703