Learn More
Aviva Systems Biology Corp HOXA11 Antibody - C-terminal region (ARP37064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Hox-1.9, Hoxa-11ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56318