Learn More
Aviva Systems Biology Corp HOXA11 Antibody - middle region (ARP58161_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58161_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXA11. Alternate names: HOX1, HOX1IApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62253