Learn More
Aviva Systems Biology Corp HOXA3 Antibody - middle region (ARP36057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXA3. Alternate names: HOX1, HOX1E, MGC10155ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55995