Learn More
Aviva Systems Biology Corp HOXA5 Antibody - middle region (ARP31447_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31447_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human HOXA5. Alternate names: HOX1, HOX1.3, HOX1C, MGC9376ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54227