Learn More
Aviva Systems Biology Corp HOXA7 Antibody - N-terminal region (ARP32637_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32637_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXA7. Alternate names: ANTP, HOX1, HOX1A, HOX1.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54684