Learn More
Aviva Systems Biology Corp HOXA9 Antibody - middle region (ARP39932_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39932_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXA9. Alternate names: ABD-B, HOX1, HOX1.7, HOX1G, MGC1934ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57237