Learn More
Aviva Systems Biology Corp HOXB1 Antibody - N-terminal region (ARP36888_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36888_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse HOXB1. Alternate names: Hox-2.9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56290