Learn More
Aviva Systems Biology Corp HOXB1 Antibody - C-terminal region (ARP38114_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38114_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HOXB1. Alternate names: HOX2, HOX2I, Hox-2.9, MGC116843, MGC116844, MGC116845ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-717