Learn More
Aviva Systems Biology Corp HOXB2 Antibody - N-terminal region (ARP35647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXB2. Alternate names: HOX2, HOX2H, Hox-2.8, K8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53016