Learn More
Aviva Systems Biology Corp HOXB3 Antibody - N-terminal region (ARP31197_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31197_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXB3. Alternate names: HOX2, HOX2G, Hox-2.7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53924