Learn More
Aviva Systems Biology Corp HOXB4 Antibody - N-terminal region (ARP39581_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39581_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXB4. Alternate names: HOX-2.6, HOX2, HOX2FApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-57092