Learn More
Aviva Systems Biology Corp HOXB5 Antibody - N-terminal region (ARP31444_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31444_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HOXB5. Alternate names: HHO.C10, HOX2, HOX2A, HU-1, Hox2.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54226