Learn More
Aviva Systems Biology Corp HOXB6 Antibody - N-terminal region (ARP31388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXB6. Alternate names: HOX2, HU-2, HOX2B, Hox-2.2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54203