Learn More
Aviva Systems Biology Corp HOXB7 Antibody - middle region (ARP37068_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37068_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse HOXB7. Alternate names: Hox-2.3, AI325018ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56319