Learn More
Aviva Systems Biology Corp Hoxb7 Antibody - middle region (ARP37068_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37068_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hoxb7. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI325018, Hox-2.3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-578