Learn More
Aviva Systems Biology Corp Hoxb9 Antibody - N-terminal region (ARP36893_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36893_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hoxb9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of mouse Hoxb9. Alternate names: Hox-2.5, MGC124051, MGC124052, MGC124053, MGC124054ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-536