Learn More
Aviva Systems Biology Corp HOXB9 Antibody - N-terminal region (ARP32639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXB9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HOXB9. Alternate names: HGNC:5120, HOX-2.5, HOX2, HOX2EApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54055