Learn More
Aviva Systems Biology Corp HOXC10 Antibody - N-terminal region (ARP39274_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC10. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HOXC10. Alternate names: HOX3I, MGC5259ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56966