Learn More
Aviva Systems Biology Corp HOXC11 Antibody - middle region (ARP32642_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32642_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HOXC11. Alternate names: HOX3H, MGC4906ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52868