Learn More
Aviva Systems Biology Corp Hoxc11 Antibody - C-terminal region (ARP39062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hoxc11. It was validated on Western Blot. . Alternate names: Hox-3.7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-779