Learn More
Aviva Systems Biology Corp HOXC12 Antibody - C-terminal region (ARP40006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HOXC12. Alternate names: HOC3F, HOX3, HOX3FApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-847