Learn More
Aviva Systems Biology Corp HOXC5 Antibody - N-terminal region (ARP39378_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39378_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXC5. Alternate names: CP11, HOX3, HOX3DApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54057