Learn More
Aviva Systems Biology Corp HOXC6 Antibody - C-terminal region (ARP38484_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38484_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HOXC6. Alternate names: CP25, HHO.C8, HOX3, HOX3CApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-737