Learn More
Aviva Systems Biology Corp HOXC8 Antibody - N-terminal region (ARP39557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOXC8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOXC8. Alternate names: HOX3, HOX3AApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57084